missing translation for 'onlineSavingsMsg'
Learn More

Dextran sulfate sodium salt, MW ca >500,000

Catalog No. p-7042508
Change view
Click to view available options
Quantity:
5 g
50 g
100 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6278706 5 g
AAJ6278718 50 g
AAJ6278722 100 g
AAJ62787A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAJ6278706 Supplier Thermo Scientific Chemicals Supplier No. J6278706
Only null left

CAS: 9011-18-1 | (C6H7O5)m(C6H7O5)n | 40,000 g/mol

Dextran sulfate sodium is used to study colitis (inflammatory bowel disease) as well as an antiviral agent. It functions as a nonsurfactant, suspending agent with gel forming and viscosity controlling properties. It can be used in gels and moisturizers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9011-18-1
Molecular Formula (C6H7O5)m(C6H7O5)n
Molecular Weight (g/mol) 40,000
MDL Number MFCD00081551
InChI Key BLFLLBZGZJTVJG-UHFFFAOYSA-N
Synonym benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
PubChem CID 2337
ChEBI CHEBI:116735
SMILES *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*]

Specifications

Color White to Yellow
Physical Form Powder
Merck Index 14,2951
Solubility Information Soluble in water.
Quantity 5 g
IUPAC Name Dextran sulfate sodium salt
Formula Weight >≈500 kd
Odor Phenol-like
Chemical Name or Material Dextran sulfate sodium salt
RTECSNumber HH9290000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only