missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextran sulfate sodium salt, approx. M.W. 6,500 - 10,000
Dextran sulfate sodium salt, C9H11NO2, CAS Number-9011-18-1, americaine, anaesthesin, anesthesin, benzocaine, ethoform, ethyl aminobenzoate, ethyl p-aminobenzoate, norcaine, orthesin, parathesin, 25g, CHEBI:116735, BLFLLBZGZJTVJG-UHFFFAOYSA-N, ethyl 4-aminobenzoate, MFCD00081551, 165.192 | CAS: 9011-18-1 | (C6H7O5)m(C6H7O5)n | 40,000 g/mol
$193.38 - $193.38
Chemical Identifiers
| CAS | 9011-18-1 |
|---|---|
| Molecular Formula | (C6H7O5)m(C6H7O5)n |
| Molecular Weight (g/mol) | 40,000 |
| MDL Number | MFCD00081551 |
| InChI Key | BLFLLBZGZJTVJG-UHFFFAOYSA-N |
| Synonym | benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin |
| PubChem CID | 2337 |
| ChEBI | CHEBI:116735 |
| SMILES | *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC441490050
|
Thermo Scientific Chemicals
441490050 |
5 g |
Each for $193.38
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 9011-18-1 | |
| 40,000 | |
| BLFLLBZGZJTVJG-UHFFFAOYSA-N | |
| 2337 | |
| *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*] |
| (C6H7O5)m(C6H7O5)n | |
| MFCD00081551 | |
| benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin | |
| CHEBI:116735 |
Specifications
| 9011-18-1 | |
| MFCD00081551 | |
| Solubility in water: 100g/L (25°C). | |
| BLFLLBZGZJTVJG-UHFFFAOYSA-N | |
| Dextran sulfate sodium salt | |
| 2337 | |
| Glass bottle |
| (C6H7O5)m(C6H7O5)n | |
| benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin | |
| 5 g | |
| *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*] | |
| 40,000 | |
| CHEBI:116735 | |
| Dextran sulfate sodium salt, approx. M.W. 6, 500 - 10, 000 |
RUO – Research Use Only