missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Dextran Sulfate Sodium Salt

 

Supplier:  Thermo Scientific™ 611361000

Quantity 100g

Catalog No. AC611361000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

9011-18-1
165.192
BLFLLBZGZJTVJG-UHFFFAOYSA-N
2337
ethyl 4-aminobenzoate
C9H11NO2
MFCD00081551
benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
CHEBI:116735
CCOC(=O)C1=CC=C(C=C1)N

Specifications

Dextran sulfate sodium salt
C9H11NO2
15, 2953
100g
CCOC(=O)C1=CC=C(C=C1)N
165.192
CHEBI:116735
7.5% max.
9011-18-1
MFCD00081551
benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
BLFLLBZGZJTVJG-UHFFFAOYSA-N
ethyl 4-aminobenzoate
2337
Pass Test
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions