missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextran sulfate sodium salt (400,000-600,000 M.Wt), MP Biomedicals™
$74.69 - $591.26
Chemical Identifiers
| CAS | 9011-18-1 |
|---|---|
| Molecular Formula | (C6H7O5)m(C6H7O5)n |
| Molecular Weight (g/mol) | 40,000 |
| MDL Number | MFCD00081551 |
| InChI Key | BLFLLBZGZJTVJG-UHFFFAOYSA-N |
| Synonym | benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin |
| PubChem CID | 2337 |
| ChEBI | CHEBI:116735 |
| SMILES | *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN16011101
|
MP Biomedicals Inc
0216011101 |
1 g |
Each for $74.69
|
|
|||||
|
ICN16011110
|
MP Biomedicals Inc
0216011110 |
10 g |
Each for $118.63
|
|
|||||
|
ICN16011150
|
MP Biomedicals Inc
0216011150 |
50 g |
Each for $361.75
|
|
|||||
|
ICN16011180
|
MP Biomedicals Inc
0216011180 |
100 g |
Each for $591.26
|
|
|||||
Chemical Identifiers
| 9011-18-1 | |
| 40,000 | |
| BLFLLBZGZJTVJG-UHFFFAOYSA-N | |
| 2337 | |
| *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*] |
| (C6H7O5)m(C6H7O5)n | |
| MFCD00081551 | |
| benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin | |
| CHEBI:116735 |
Specifications
| White | |
| 9011-18-1 | |
| MFCD00081551 | |
| 1 g | |
| *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*] | |
| 40,000 | |
| CHEBI:116735 | |
| Dextran Sulfate Sodium Salt |
| Powder | |
| (C6H7O5)m(C6H7O5)n | |
| benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin | |
| BLFLLBZGZJTVJG-UHFFFAOYSA-N | |
| Dextran sulfate sodium salt | |
| 2337 | |
| 400,000 to 600,000 |