missing translation for 'onlineSavingsMsg'
Learn More

Dextran sulfate sodium salt (400,000-600,000 M.Wt), MP Biomedicals™

Catalog No. ICN16011101
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN16011101 1 g
ICN16011110 10 g
ICN16011150 50 g
ICN16011180 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN16011101 Supplier MP Biomedicals Inc Supplier No. 0216011101

Chemical Identifiers

CAS 9011-18-1
Molecular Formula (C6H7O5)m(C6H7O5)n
Molecular Weight (g/mol) 40,000
MDL Number MFCD00081551
InChI Key BLFLLBZGZJTVJG-UHFFFAOYSA-N
Synonym benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
PubChem CID 2337
ChEBI CHEBI:116735
SMILES *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*]

Specifications

Color White
Physical Form Powder
Quantity 1 g
IUPAC Name Dextran sulfate sodium salt
Formula Weight 400,000 to 600,000
Chemical Name or Material Dextran Sulfate Sodium Salt