missing translation for 'onlineSavingsMsg'
Learn More

Dextran Pyrogen Free Clinical MP Biomedicals

Supplier:  MP Biomedicals Inc 0210151491

 View more versions of this product

Catalog No. ICN10151491

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Dextran
9004-54-0
MFCD00130935
1 kg
O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O
504.44
200,000 to 300,000
Pyrogen Free
(C6H9O5)n(C6H10O5)m
dextran, dextran, mw-86.000 aver., 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy methyl tetrahydro-2h-pyran-2-yl oxy hexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxyhexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxymethyl tetrahydropyran-2-yl oxy-hexanal
FZWBNHMXJMCXLU-UHFFFAOYNA-N
2,3,4,5-tetrahydroxy-6-[3,4,5-trihydroxy-6-[[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxymethyl]oxan-2-yl]oxyhexanal
4125253
Clinical

Chemical Identifiers

9004-54-0
504.44
FZWBNHMXJMCXLU-UHFFFAOYNA-N
4125253
(C6H9O5)n(C6H10O5)m
MFCD00130935
dextran, dextran, mw-86.000 aver., 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy methyl tetrahydro-2h-pyran-2-yl oxy hexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxyhexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxymethyl tetrahydropyran-2-yl oxy-hexanal
O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions