Learn More
Dextran, MW ca 75,000
Dextran, CAS # 9004-54-0, is a glucose polymer. It is mainly used to increase electronegativity and reduce erythrocyte aggregation and platelet adhesiveness (anticoagulant activity). | CAS: 9004-54-0 | (C6H9O5)n(C6H10O5)m | 504.44 g/mol
Supplier: Thermo Scientific Chemicals J6098918
Description
Dextran, MW ca 75,000 is used as a processing aid, stabilizer, thickener, and surface finishing agent.. Further, it is used in soft center confections. In addition to this, it is used as a barley malt substitute and as a plasma volume expander.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Dextran is a glucose polymer biosynthesized by the non-pathogenic Leuconostoc mesenteroides
Application
• Dextran is a complex branched glucan that can be used to enhance electronegativity and reduce erythrocyte aggregation and platelet adhesiveness (anticoagulant activity)
• This compound promotes the activation of plasminogen by inhibiting α-2 antiplasmin
• Dextran can be used to reduce thrombosis in animal models
• It is used in osmotic stress techniques where osmotic pressure is applied to biological molecules • Dextran can be used as an immobilization agent in biosensors
Specifications
| White | |
| Powder | |
| (C6H9O5)n(C6H10O5)m | |
| 14,2948 | |
| Soluble in water,dimethyl formamide and glycerol. | |
| FZWBNHMXJMCXLU-UHFFFAOYNA-N | |
| Dextran | |
| 4125253 |
| Dextran | |
| 9004-54-0 | |
| MFCD00130935 | |
| dextran, dextran, mw-86.000 aver., 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy methyl tetrahydro-2h-pyran-2-yl oxy hexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxyhexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxymethyl tetrahydropyran-2-yl oxy-hexanal | |
| 50 g | |
| O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O | |
| 504.44 | |
| ≈75 kd |
Chemical Identifiers
| 9004-54-0 | |
| 504.44 | |
| FZWBNHMXJMCXLU-UHFFFAOYNA-N | |
| 4125253 | |
| O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O |
| (C6H9O5)n(C6H10O5)m | |
| MFCD00130935 | |
| dextran, dextran, mw-86.000 aver., 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy methyl tetrahydro-2h-pyran-2-yl oxy hexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxyhexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxymethyl tetrahydropyran-2-yl oxy-hexanal | |
| Dextran |
Safety and Handling
EINECSNumber : 232-677-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only