missing translation for 'onlineSavingsMsg'
Learn More

Dextran, MW ca 6,000

Catalog No. AAJ6277509
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6277509 10 g
AAJ6277518 50 g
AAJ6277522 100 g
AAJ6277536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAJ6277509 Supplier Thermo Scientific Chemicals Supplier No. J6277509

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Dextran, also known as manninotriose, CAS # 9004-54-0, is a glucose polymer of bacterial origin. | CAS: 9004-54-0 | (C6H9O5)n(C6H10O5)m | 504.44 g/mol

Dextran is used in volume extenders, stabilizers, matrix components, binding platforms, lubricants and physical structure components.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Dextran is a complex branched polysaccharide of bacterial origin
• It primarily contains alpha-D C-1 – C-6 glycosidic linkages
• Alternate linkages account for the complex branching of dextran

Application

• Some research suggests that dextran can be used a carrier that helps target delivery of compounds to specific cells
• It is intended for protein isolation and centrifugation protocols, as well as other research applications

Chemical Identifiers

CAS 9004-54-0
Molecular Formula (C6H9O5)n(C6H10O5)m
Molecular Weight (g/mol) 504.44
MDL Number MFCD00130935
InChI Key FZWBNHMXJMCXLU-UHFFFAOYNA-N
Synonym dextran, dextran, mw-86.000 aver., 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy methyl tetrahydro-2h-pyran-2-yl oxy hexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxyhexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxymethyl tetrahydropyran-2-yl oxy-hexanal
PubChem CID 4125253
IUPAC Name Dextran
SMILES O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O

Specifications

Color White
Physical Form Powder
Quantity 10 g
Merck Index 14,2948
Solubility Information Soluble in water,dimethyl sulfoxide,ethylene glycol and glycerol.
Formula Weight ≈6 kd
Chemical Name or Material Dextran
EINECSNumber 232-677-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only