missing translation for 'onlineSavingsMsg'
Learn More

Dextran, MW ca 250,000

Catalog No. p-7041773
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6020009 10 g
AAJ6020018 50 g
AAJ6020022 100 g
AAJ6020036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAJ6020009 Supplier Thermo Scientific Chemicals Supplier No. J6020009
Only null left

CAS: 9004-54-0 | (C6H9O5)n(C6H10O5)m | 504.44 g/mol

Dextran, is used as a multi-use, branched glucan. Low molecular weight dextran is useful as a stabilizer, volume extender, lubricant, binding platform, and matrix component. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9004-54-0
Molecular Formula (C6H9O5)n(C6H10O5)m
Molecular Weight (g/mol) 504.44
MDL Number MFCD00130935
InChI Key FZWBNHMXJMCXLU-UHFFFAOYNA-N
Synonym dextran, dextran, mw-86.000 aver., 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy methyl tetrahydro-2h-pyran-2-yl oxy hexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxyhexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxymethyl tetrahydropyran-2-yl oxy-hexanal
PubChem CID 4125253
IUPAC Name Dextran
SMILES O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O

Specifications

Color White
Physical Form Powder
Merck Index 14,2948
Solubility Information It is soluble in water (100mg/ml),DMSO (freely),formamide (freely),ethylene glycol (freely),and glycerol (freely).
Quantity 10 g
Formula Weight ≈250 kd
Chemical Name or Material Dextran
EINECSNumber 232-677-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only