missing translation for 'onlineSavingsMsg'
Learn More

Dextran, MW ca 150,000

Dextran, CAS # 9004-54-0, is a glucose polymer. It is mainly used to increase electronegativity and reduce erythrocyte aggregation and platelet adhesiveness (anticoagulant activity). | CAS: 9004-54-0 | (C6H9O5)n(C6H10O5)m | 504.44 g/mol

Supplier:  Thermo Scientific Chemicals J6378936

 View more versions of this product

Catalog No. AAJ6378936


Only null left
Explore available promotions

Description

Description

Dextran is used in a bead form to aid in bioreactor applications, some size-exclusion chromatography matrices and in osmotic stress technique study involved in biological molecules. It can be used as a stabilizing coating to protect metal nanoparticles from oxidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Dextran is a glucose polymer biosynthesized by the non-pathogenic Leuconostoc mesenteroides

Application

• Dextran is a complex branched glucan that can be used to enhance electronegativity and reduce erythrocyte aggregation and platelet adhesiveness (anticoagulant activity)
• This compound promotes the activation of plasminogen by inhibiting α-2 antiplasmin
• Dextran can be used to reduce thrombosis in animal models
• It is used in osmotic stress techniques where osmotic pressure is applied to biological molecules
• Dextran can be used as an immobilization agent in biosensors

Specifications

Specifications

White
Powder
(C6H9O5)n(C6H10O5)m
14,2948
Soluble in water,dimethyl sulfoxide,ethylene glycol and glycerol.
FZWBNHMXJMCXLU-UHFFFAOYNA-N
Dextran
4125253
Dextran
9004-54-0
MFCD00130935
dextran, dextran, mw-86.000 aver., 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy methyl tetrahydro-2h-pyran-2-yl oxy hexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxyhexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxymethyl tetrahydropyran-2-yl oxy-hexanal
500 g
O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O
504.44
≈150 kd

Chemical Identifiers

9004-54-0
504.44
FZWBNHMXJMCXLU-UHFFFAOYNA-N
4125253
O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O
(C6H9O5)n(C6H10O5)m
MFCD00130935
dextran, dextran, mw-86.000 aver., 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy methyl tetrahydro-2h-pyran-2-yl oxy hexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxyhexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxymethyl tetrahydropyran-2-yl oxy-hexanal
Dextran
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 232-677-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only