missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextran, MP Biomedicals™
Specifications
Specifications
| Color | White |
| Physical Form | Powder |
| Quantity | 10 g |
| Formula Weight | 35000 to 50000 |
| Chemical Name or Material | Dextran |
Safety and Handling
| Recommended Storage | Store at Room Temperature (15° to 30°C) |