missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextran, MP Biomedicals™
Specifications
Specifications
| Quantity | 10 g |
| IUPAC Name | Dextran |
| Formula Weight | 40000 |
| Chemical Name or Material | Dextran |
Safety and Handling
| Recommended Storage | Store at Room Temperature (15° to 30°C) |