missing translation for 'onlineSavingsMsg'
Learn More

Dextran, for GPC 270,000, MilliporeSigma™ Supelco™

Catalog No. p-7187490
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015808 100 mg
111015822 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015808 Supplier Merck Emd Millipore Supplier No. 00894100MG
Only null left

Dextran is a polysaccharide. Dextran fluids behave as colloids; and also are used as plasma substitution. It may have siliconizing effect helping in coating raw serosal surfaces.

Featured Industries: Food and Beverages

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 9004-54-0
Physical Form Powder/Crystals
Linear Formula [C6H10O5 ]n
Technique GPC
MDL Number MFCD00130935
Type Analytical Standard
Quantity 100 mg
Grade Analytical Standard