missing translation for 'onlineSavingsMsg'
Learn More

Dextran for for GPC 150,000, MilliporeSigma™ Supelco™

Catalog No. 111015807
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015807 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015807 Supplier Merck Emd Millipore Supplier No. 00893100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Dextran is a polysaccharide. Dextran fluids behave as colloids; and also are used as plasma substitution. It may have siliconizing effect helping in coating raw serosal surfaces.

Featured Industries: Food and Beverages

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 9004-54-0
Physical Form Crystalline
Linear Formula [C6H10O5 ]n
Technique GPC
MDL Number MFCD00130935
Type Analytical Standard
Quantity 100 mg
Grade Analytical Standard