missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextran 70 (Mw.=ca. 70,000), TCI America™
Specifications
Specifications
| Color | White |
| Physical Form | Crystal-Powder at 20°C |
| Quantity | 25 g |
| Chemical Name or Material | Dextran 70 (Mw.=ca. 70,000) |
Safety and Handling
| EINECSNumber | (8)-0098 |
| RTECSNumber | HH9247500 |
| TSCA | Yes |