missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextran, 500,000, Powder, Spectrum™ Chemical
D1004, 9004-54-0
$294.30 - $3,667.17
Chemical Identifiers
| CAS | 9004-54-0 |
|---|---|
| Molecular Formula | (C6H9O5)n(C6H10O5)m |
| Molecular Weight (g/mol) | 504.44 |
| MDL Number | MFCD00130935 |
| InChI Key | FZWBNHMXJMCXLU-UHFFFAOYNA-N |
| IUPAC Name | Dextran |
| SMILES | O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18602090
|
Spectrum Chemical Mfg Cor
D100425GM |
25 g | Poly Bottle |
N/A
|
|
||||
|
18602091
|
Spectrum Chemical Mfg Cor
D1004100GM |
100 g | Poly Bottle |
N/A
|
|
||||
|
18602092
|
Spectrum Chemical Mfg Cor
D10041KG |
1 kg | Poly Bottle |
N/A
|
|
||||
|
18602093
|
Spectrum Chemical Mfg Cor
D100410KGBL |
10 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18602094
|
Spectrum Chemical Mfg Cor
D100450KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Dextran, 500,000, Powder is a complex branched glucan that is synthethized by certain lactic acid bacteria from sucrose. In the laboratory, Dextran is used to apply osmotic pressure to biological molecules and can, when applied with a fluorescent molecule, be used to create concentration gradients of diffusible molecules.Chemical Identifiers
| 9004-54-0 | |
| 504.44 | |
| FZWBNHMXJMCXLU-UHFFFAOYNA-N | |
| O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O |
| (C6H9O5)n(C6H10O5)m | |
| MFCD00130935 | |
| Dextran |
Specifications
| White powder | |
| 100% | |
| (C6H9O5)n(C6H10O5)m | |
| 25 g | |
| O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O | |
| 504.44 | |
| 10% |
| 9004-54-0 | |
| 2% | |
| MFCD00130935 | |
| FZWBNHMXJMCXLU-UHFFFAOYNA-N | |
| Dextran | |
| Reagent | |
| Poly Bottle |