missing translation for 'onlineSavingsMsg'
Learn More

Dextran, 40,000, Powder, Spectrum™ Chemical

Catalog No. 18602089
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
25 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602089 25 kg Poly Bottle
18602086 25 g Poly Bottle
18602087 100 g Poly Bottle
18602088 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18602089 Supplier Spectrum Chemical Mfg Cor Supplier No. DE12425KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

DE124, 9004-54-0

Spectrum™ Chemical Dextran, 40,000, Powder is a complex branched glucan that is synthesized by certain lactic acid bacteria from sucrose. Commonly used in microsurgery to decrease the risk of vascular thrombosis, these agents are also used as effective osmotic agents and can treat hypovolemia. In the laboratory, Dextran is used to apply osmotic pressure to biological molecules and can, when applied with a fluorescent molecule, be used to create concentration gradients of diffusible molecules.

Chemical Identifiers

CAS 9004-54-0
Molecular Formula (C6H9O5)n(C6H10O5)m
Molecular Weight (g/mol) 504.44
MDL Number MFCD00130935
InChI Key FZWBNHMXJMCXLU-UHFFFAOYNA-N
IUPAC Name Dextran
SMILES O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O

Specifications

Color White powder
CAS Max % 100%
Quantity 25 kg
Loss on Drying 12%
Packaging Poly Bottle
Specific Rotation +192° to +203°
Grade Ungraded
Chemical Name or Material Dextran