missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextran, 250,000, Powder, Spectrum™ Chemical
DE130, 9004-54-0
$334.39 - $4225.64
Chemical Identifiers
| CAS | 9004-54-0 |
|---|---|
| Molecular Formula | (C6H9O5)n(C6H10O5)m |
| Molecular Weight (g/mol) | 504.44 |
| MDL Number | MFCD00130935 |
| InChI Key | FZWBNHMXJMCXLU-UHFFFAOYNA-N |
| IUPAC Name | Dextran |
| SMILES | O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18602083
|
Spectrum Chemical Mfg Cor
DE13025GM |
25 g | Poly Bottle |
Each for $334.39
|
|
||||
|
18602084
|
Spectrum Chemical Mfg Cor
DE130100GM |
100 g | Poly Bottle |
Each for $809.11
|
|
||||
|
18602085
|
Spectrum Chemical Mfg Cor
DE1301KG |
1 kg | Poly Bottle |
Each for $4,225.64
|
|
||||
Description
Spectrum™ Chemical Dextran, 250,000, Powder is a complex branched glucan that is synthesized by certain lactic acid bacteria from sucrose. Commonly used in microsurgery to decrease the risk of vascular thrombosis, these agents are also used as effective osmotic agents and can treat hypovolemia. In the laboratory, Dextran is used to apply osmotic pressure to biological molecules and can, when applied with a fluorescent molecule, be used to create concentration gradients of diffusible molecules.Chemical Identifiers
| 9004-54-0 | |
| 504.44 | |
| FZWBNHMXJMCXLU-UHFFFAOYNA-N | |
| O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O |
| (C6H9O5)n(C6H10O5)m | |
| MFCD00130935 | |
| Dextran |
Specifications
| White | |
| 100% | |
| (C6H9O5)n(C6H10O5)m | |
| 25 g | |
| O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O | |
| 504.44 | |
| 10% |
| 9004-54-0 | |
| 2% | |
| MFCD00130935 | |
| FZWBNHMXJMCXLU-UHFFFAOYNA-N | |
| Dextran | |
| Reagent | |
| Poly Bottle |