missing translation for 'onlineSavingsMsg'
Learn More

Dextran 150,000, Powder, Spectrum™ Chemical

Catalog No. 18610141
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610141 25 g Poly Bottle
18610140 100 g Poly Bottle
18610139 1 kg Poly Pail
18610142 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18610141 Supplier Spectrum Chemical Mfg Cor Supplier No. D101325GM
Only null left

Chemical Identifiers

CAS 9004-54-0
Molecular Formula (C6H9O5)n(C6H10O5)m
MDL Number MFCD00130935
IUPAC Name Dextran
SMILES O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O

Specifications

Color White to Slightly Off White
pH 4.0 to 7.0
CAS Max % 1
Quantity 25 g
Grade Reagent
Loss on Drying 0.1
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.