missing translation for 'onlineSavingsMsg'
Learn More

Dexpanthenol Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015605
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015605 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015605 Supplier Merck Emd Millipore Supplier No. PHR1228500MG
Dexpanthenol is an alcohol analog of pantothenic acid and an important component of parenterals, local cosmetic preparations and multi-vitamins. It has as much biological activity as pantothenic acid and is found to be stable in aqueous solution.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule), Vitamins, Nutraceuticals and Natural Products

Specifications

CAS 81-13-0
Density 1.20 g/mL (at 20°C)
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C9H19 NO4
MDL Number MFCD00065006
Synonym D-Panthenol; (R)-(+)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutyramide; (R)-2,4-Dihydroxy-3,3-dimethylbutyric 3-hydroxypropylamide; D-Pantothenyl alcohol; Dexpanthenol; Provitamin B
Type Certified Reference Material
Quantity 500 mg
Formula Weight 205.25
Grade Certified Reference Material
Show More Show Less