missing translation for 'onlineSavingsMsg'
Learn More

Dexmedetomidine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7128636
Change view
Click to view available options
Quantity:
20 mg
100 mg
Catalog No. Quantity
D506220MG 20 mg
D5062100MG 100 mg
2 options

Catalog No. D506220MG

Supplier: TCI America D506220MG

Only null left

Chemical Identifiers

CAS 145108-58-3
Molecular Formula C13H17ClN2
Molecular Weight (g/mol) 236.743
MDL Number MFCD12912677
InChI Key VPNGEIHDPSLNMU-MERQFXBCSA-N
Synonym dexmedetomidine hydrochloride, dexmedetomidine hcl, precedex, 4-s-alpha,2,3-trimethylbenzyl imidazole monohydrochloride, unii-1018wh7f9i, dexmedetomidine hcl precedex, s-5-1-2,3-dimethylphenyl ethyl-1h-imidazole hydrochloride, dexdor, 4-1s-1-2,3-dimethylphenyl ethyl-1h-imidazole hydrochloride
PubChem CID 6918081
ChEBI CHEBI:31472
IUPAC Name 5-[(1S)-1-(2,3-dimethylphenyl)ethyl]-1H-imidazole;hydrochloride
SMILES CC1=C(C(=CC=C1)C(C)C2=CN=CN2)C.Cl

Specifications

Melting Point 158°C
Color White
Quantity 20 mg
Physical Form Crystal-Powder at 20°C
Formula Weight 236.74
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material Dexmedetomidine Hydrochloride
RTECSNumber NI5156750
TSCA No
Recommended Storage Freezer