missing translation for 'onlineSavingsMsg'
Learn More

Dexamethasone Acetate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012681
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012681 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012681 Supplier Merck Emd Millipore Supplier No. PHR15271G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Dexamethasone Acetate is a steroid used for the treatment of various diseases because of its anti-inflammatory and inmunosuppresor effects. It is a common component of ointments, creams, lotions, aerosols and microemulsions. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 1177-87-3
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C24H31 FO6
MDL Number MFCD00027407
Synonym Dexamethasone 21-acetate; 9 alpha-Fluoro-16 alpha-methyl-11 beta,17 alpha,21-trihydroxy-1,4-pregnadiene-3,20-dione-21-acetate; 9 alpha-Fluoro-16 alpha-methylprednisolone-21-acetate; Prednisolone F acetate
Type Certified Reference Material
Quantity 1 g
Formula Weight 434.50
Grade Certified Reference Material