missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Devarda's Alloy, Granular, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 125 g |
| Packaging | Poly Bottle |
| Form | Powder |
| Grade | Reagent |
| Chemical Name or Material | Devarda's Alloy |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.