missing translation for 'onlineSavingsMsg'
Learn More

Devarda's Alloy, Granular, Spectrum™ Chemical

Catalog No. 18610136
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610136 125 g Poly Bottle
18610138 500 g Poly Bottle
18610137 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18610136 Supplier Spectrum Chemical Mfg Cor Supplier No. D1005125GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 8049-11-4

Specifications

CAS Max % 1
Quantity 125 g
Packaging Poly Bottle
Form Powder
Grade Reagent
Chemical Name or Material Devarda's Alloy

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.