missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Detritylation Reagent, Honeywell Burdick & Jackson™
BioSyn™, suitable for use with DNA/RNA synthesis instruments, 3% Dichloroacetic Acid in Toluene (v/v)
$2499.73 - $2499.73
Specifications
| CAS | 79-43-6,108-88-3 |
|---|---|
| CAS Max % | 3.00%,97.00% |
| Physical Form | Liquid, clear |
| Purity Grade Notes | Deblocking reagents |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
60-048-416
|
Honeywell Inc
SR6744 |
4 L |
Case of 4 Each for $2,499.73
|
|
|||||
Specifications
| 79-43-6,108-88-3 | |
| 3.00%,97.00% | |
| Liquid, clear | |
| Deblocking reagents |