missing translation for 'onlineSavingsMsg'
Learn More

Detritylation Reagent, Solstice

Catalog No. 60048416
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60048416 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60048416 Supplier Honeywell Inc Supplier No. SR6744

missing translation for 'validMainframeMsg'

BioSyn™, suitable for use with DNA/RNA synthesis instruments, 3% Dichloroacetic Acid in Toluene (v/v)

Specifications

CAS 79-43-6, 108-88-3
CAS Max % 3.00%, 97.00%
Quantity 4 L
Flash Point 4°C.Toluene
Packaging Stainless Steel NOWPAK™
Physical Form Liquid, clear
Purity Grade Notes Deblocking reagents