missing translation for 'onlineSavingsMsg'
Learn More

Detritylation Reagent, Honeywell Burdick & Jackson™

BioSyn™, suitable for use with DNA/RNA synthesis instruments, 3% Dichloroacetic Acid in Dichloromethane (v/v)

$2,260.32 - $2,260.32

Specifications

CAS 79-43-6,75-09-2
CAS Min % 2.90%,96.50%
CAS Max % 3.50%,97.10%
Boiling Point 40°C.The physical data is that of the main component.
Flash Point 98.9°C.The physical data is that of the main component.
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
60-048-418
View Documents
Honeywell Inc
SR6224X4L
4 L N/A
Only null left
 
Description

Description

Specifications

Specifications

79-43-6,75-09-2
3.50%,97.10%
98.9°C.The physical data is that of the main component.
Liquid, clear
2.90%,96.50%
40°C.The physical data is that of the main component.
Returnable NOWPAK™ II
Deblocking reagents
Videos
SDS
Documents

Documents