missing translation for 'onlineSavingsMsg'
Learn More

Detritylation Reagent, Solstice

Catalog No. 60048418
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60048418 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60048418 Supplier Honeywell Inc Supplier No. SR6224X4L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BioSyn™, suitable for use with DNA/RNA synthesis instruments, 3% Dichloroacetic Acid in Dichloromethane (v/v)

Specifications

CAS 79-43-6, 75-09-2
CAS Min % 2.90%, 96.50%
CAS Max % 3.50%, 97.10%
Quantity 4 L
Flash Point 98.9°C.The physical data is that of the main component.
Packaging Returnable NOWPAK™ II
Physical Form Liquid, clear
Purity Grade Notes Deblocking reagents
Boiling Point 40°C.The physical data is that of the main component.