Learn More
Azure Biosystems Dessicant Replacement Container
Azure Biosytems Dessicant Replacement Container is a replacement part for the chemiSOLO imaging system's camera dessicant.
Supplier: Azure Biosystems CS1021
Description
- Recommended for replacement after one year of regular chemiSOLO imaging
Specifications
| Upgrade Kit |