missing translation for 'onlineSavingsMsg'
Learn More

Dess-Martin Periodinane 95.0+%, TCI America™

Supplier:  TCI America D20455G

 View more versions of this product

Catalog No. D20455G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Dess-Martin Periodinane
124°C
C13H13IO8
5 g
dess-martin periodinane, triacetoxyperiodinane, 1,1,1-triacetoxy-1,1-dihydro-1,2-benziodoxol-3 1h-one, dess-martin reagent, dess-martinperiodinane, dess-martin, dess martin periodinane, dess martin, dess-martin periodane
CC(=O)O[I]1(OC(C)=O)(OC(C)=O)OC(=O)C2=CC=CC=C12
424.14
424.14
Crystalline Powder
87413-09-0
White-Yellow
MFCD00130127
1479
NKLCNNUWBJBICK-UHFFFAOYSA-N
1,1-bis(acetyloxy)-3-oxo-3H-1λ⁵,2-benziodaoxol-1-yl acetate
159087
≥95.0% (T)

Chemical Identifiers

87413-09-0
424.14
NKLCNNUWBJBICK-UHFFFAOYSA-N
159087
CC(=O)O[I]1(OC(C)=O)(OC(C)=O)OC(=O)C2=CC=CC=C12
C13H13IO8
MFCD00130127
dess-martin periodinane, triacetoxyperiodinane, 1,1,1-triacetoxy-1,1-dihydro-1,2-benziodoxol-3 1h-one, dess-martin reagent, dess-martinperiodinane, dess-martin, dess martin periodinane, dess martin, dess-martin periodane
1,1-bis(acetyloxy)-3-oxo-3H-1λ⁵,2-benziodaoxol-1-yl acetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions