missing translation for 'onlineSavingsMsg'
Learn More

Dess-Martin Periodinane 95.0+%, TCI America™

Catalog No. p-7127083
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20451G 1 g
D20455G 5 g
D204525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D20451G Supplier TCI America Supplier No. D20451G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 87413-09-0 | C13H13IO8 | 424.14 g/mol

Chemical Identifiers

CAS 87413-09-0
Molecular Formula C13H13IO8
Molecular Weight (g/mol) 424.14
MDL Number MFCD00130127
InChI Key NKLCNNUWBJBICK-UHFFFAOYSA-N
Synonym dess-martin periodinane, triacetoxyperiodinane, 1,1,1-triacetoxy-1,1-dihydro-1,2-benziodoxol-3 1h-one, dess-martin reagent, dess-martinperiodinane, dess-martin, dess martin periodinane, dess martin, dess-martin periodane
PubChem CID 159087
IUPAC Name 1,1-bis(acetyloxy)-3-oxo-3H-1λ⁵,2-benziodaoxol-1-yl acetate
SMILES CC(=O)O[I]1(OC(C)=O)(OC(C)=O)OC(=O)C2=CC=CC=C12

Specifications

Melting Point 124°C
Color White-Yellow
Quantity 1 g
UN Number 1479
Formula Weight 424.14
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dess-Martin Periodinane
TSCA No
Recommended Storage Freezer