missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Desmoglein 2 Recombinant Rabbit Monoclonal Antibody (23GB5215)
Rabbit Recombinant Monoclonal Antibody
$656.94 - $1779.35
Specifications
| Antigen | Desmoglein 2 |
|---|---|
| Clone | 23GB5215 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
Description
Desmoglein 2 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
Desmosomes are cell-cell junctions between epithelial, myocardial, and certain other cell types. Currently, three desmoglein subfamily members have been identified and all are members of the cadherin cell adhesion molecule superfamily. These desmoglein gene family members are located in a cluster on chromosome 18. This protein has been identified as the autoantigen of the autoimmune skin blistering disease pemphigus vulgaris.Specifications
| Desmoglein 2 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| DSG2 | |
| AA408168; ARVC10; ARVD10; Cadherin family member 5; CDHF5; CMD1BB; D18Ertd293e; desmoglein 2; desmoglein-2; DSG2; DSG-2; HDGC; MGC117034; MGC117036; MGC117037 | |
| DSG2 | |
| Primary | |
| Affinity chromatography |
| 23GB5215 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q14126 | |
| 1829 | |
| A synthesized peptide derived from human Desmoglein 2 (1050-1118AA). | |
| Antibody |