missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Desmoglein 2 Monoclonal Antibody (10D2)
GREENER_CHOICE

Catalog No. PIMA536301
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536301 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536301 Supplier Invitrogen™ Supplier No. MA536301
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

Tag sequence is: LLPKHPHLVR QKRAWITAPV ALREGEDLSK KNPIAKIHSD LAEERGLKIT YKYTGKGITE PPFGIFVFN KDTGELNVTS ILDREETPFF LLTGYALDAR GNNVEKPLEL RIKVLDIND NEPVFTQDVF Immunogen sequence: LLPKHPHLVR QKRAWITAPV ALREGEDLSK KNPIAKIHSD LAEERGLKIT YKYTGKGITE PPFGIFVFNK DTGELNVTSI LDREETPFFL LTGYALDARGN NVEKPLELRI KVLDINDNEP VFTQDVF.

Desmosomes are cell-cell junctions between epithelial, myocardial, and certain other cell types. Currently, three desmoglein subfamily members have been identified and all are members of the cadherin cell adhesion molecule superfamily. These desmoglein gene family members are located in a cluster on chromosome 18. This protein has been identified as the autoantigen of the autoimmune skin blistering disease pemphigus vulgaris.

Specifications

Antigen Desmoglein 2
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 10D2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene DSG2
Gene Accession No. Q14126
Gene Alias AA408168; ARVC10; ARVD10; Cadherin family member 5; CDHF5; CMD1BB; D18Ertd293e; desmoglein 2; desmoglein-2; DSG2; DSG-2; HDGC; MGC117034; MGC117036; MGC117037
Gene Symbols DSG2
Host Species Mouse
Immunogen N terminal amino acids 37-164 of Human Desmoglein 2 fused to a proprietary tag.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1829
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less