missing translation for 'onlineSavingsMsg'
Learn More

Desloratadine 98.0+%, TCI America™

Catalog No. D3787100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3787100MG 100 mg
D37871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3787100MG Supplier TCI America Supplier No. D3787100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 100643-71-8
Molecular Formula C19H19ClN2
Molecular Weight (g/mol) 310.825
MDL Number MFCD00871949
InChI Key JAUOIFJMECXRGI-UHFFFAOYSA-N
PubChem CID 124087
ChEBI CHEBI:291342
IUPAC Name 8-chloro-11-piperidin-4-ylidene-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridine
SMILES C1CC2=C(C=CC(=C2)Cl)C(=C3CCNCC3)C4=C1C=CC=N4

Specifications

Melting Point 157°C
Quantity 100 mg
Formula Weight 310.83
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Desloratadine
RTECSNumber DE8011000
TSCA No
Recommended Storage Refrigerator