missing translation for 'onlineSavingsMsg'
Learn More

Desbenzyl donepezil hydrochloride, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111012739
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012739 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012739 Supplier Merck Emd Millipore Supplier No. PHR202550MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Desbenzyl donepezil hydrochloride is a metabolite of drug, donepezil hydrochloride. Donepezil hydrochloride is a neurocognition-enhancing drug prescribed especially to manage dementia during Alzheimer′s disease.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

CAS 120013-39-0
Linear Formula C17H23NO3 · HCl
Synonym 5,6-Dimethoxy-2-(piperidin-4-ylmethyl)indan-1-one hydrochloride
Quantity 50 mg
Formula Weight 325.83
Grade Certified Reference Material