missing translation for 'onlineSavingsMsg'
Learn More

DEPC-Treated Water, MP Biomedicals™

Catalog No. MP112420204
Change view
Click to view available options
Quantity:
5 x 200 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP112420204 5 x 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP112420204 Supplier MP Biomedicals Inc Supplier No. 112420204

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Formula Weight 18.02
Quantity 5 x 200 mL
Chemical Name or Material Water