missing translation for 'onlineSavingsMsg'
Learn More

Deoxyribonucleic acid sodium salt, salmon testes

Catalog No. AAJ6575306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6575306 5 g
AAJ6575303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6575306 Supplier Thermo Scientific Chemicals Supplier No. J6575306

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 68938-01-2 | C17H15BrFNO3 | 50000-100000 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 68938-01-2
Molecular Formula C17H15BrFNO3
Molecular Weight (g/mol) 50000-100000
MDL Number MFCD00130922
InChI Key GXZCYECDYOBPGH-UHFFFAOYNA-N
Synonym SS-DNA
PubChem CID 73904662
IUPAC Name (2R)-3-(3-bromophenyl)-2-[[2-(4-fluorophenyl)acetyl]amino]propanoic acid
SMILES *

Specifications

Color White
Quantity 5 g
Merck Index 14,2907
Physical Form Solid
Chemical Name or Material Deoxyribonucleic acid sodium salt, salmon testes
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only