accessibility menu, dialog, popup

UserName

A(a)

CAS RN 68938-01-2

IUPAC Name: Deoxyribonucleic acid sodium salt
Synonyms: DNA sodium salt
Molecular Weight (g/mol): —
Molecular Formula: —
InChi Key: —
SMILES: *
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results