missing translation for 'onlineSavingsMsg'
Learn More

Deoxynivalenol 3-glucoside Solution, 50 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™

Catalog No. 111018033
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018033 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018033 Supplier Merck Emd Millipore Supplier No. 329111ML
Only null left

Certan Vial

Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 131180-21-7
Flash Point 6°C - Closed Cup
Linear Formula C21H30O11
MDL Number MFCD11114484
Type Analytical Standard
Quantity 1 mL
Formula Weight 458.46
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 50 μg/mL in acetonitrile