missing translation for 'onlineSavingsMsg'
Learn More

Deoxynivalenol-13C15 Solution, 25 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™

Catalog No. 111017942
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017942 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017942 Supplier Merck Emd Millipore Supplier No. 341281ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Deoxynivalenol-13C15 (13C15-DON) is a fusarium mycotoxin which belongs to trichothecene class of compounds.

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

CAS 911392-36-4
Flash Point 2°C
Linear Formula 13C15H20O6
MDL Number MFCD11114483
Synonym 13C15 DON; Fully 13 C-labelled Deoxynivalenol
Type Analytical Standard
Quantity 1 mL
Formula Weight 311.21
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 25 μg/mL in acetonitrile