missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Deoxycholic Acid, Sodium Salt, ≥99%, ULTROL™ Grade, Calbiochem™,
$227.00 - $842.00
Chemical Identifiers
| CAS | 302-95-4 |
|---|---|
| Molecular Formula | C24H39NaO4 |
| Molecular Weight (g/mol) | 414.56 |
| MDL Number | MFCD00064139 |
| InChI Key | FHHPUSMSKHSNKW-SMOYURAASA-M |
| PubChem CID | 91810855 |
| IUPAC Name | sodium (4R)-4-[(1R,3aS,3bR,5aR,7R,9aS,9bS,11S,11aR)-7,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate |
| SMILES | [Na+].[H][C@@]12CC[C@H]([C@H](C)CCC([O-])=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])CC[C@]2([H])C[C@H](O)CC[C@]12C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
2641035GM
|
MilliporeSigma™
2641035GM |
5 g | Glass Bottle |
Each for $227.00
|
|
||||
|
26410325GM
|
MilliporeSigma™
26410325GM |
25 g | Glass Bottle |
Each for $842.00
|
|
||||
Chemical Identifiers
| 302-95-4 | |
| 414.56 | |
| FHHPUSMSKHSNKW-SMOYURAASA-M | |
| sodium (4R)-4-[(1R,3aS,3bR,5aR,7R,9aS,9bS,11S,11aR)-7,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate |
| C24H39NaO4 | |
| MFCD00064139 | |
| 91810855 | |
| [Na+].[H][C@@]12CC[C@H]([C@H](C)CCC([O-])=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])CC[C@]2([H])C[C@H](O)CC[C@]12C |
Specifications
| White | |
| C24H39NaO4 | |
| FHHPUSMSKHSNKW-SMOYURAASA-M | |
| sodium (4R)-4-[(1R,3aS,3bR,5aR,7R,9aS,9bS,11S,11aR)-7,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate | |
| 91810855 | |
| ULTROL |
| 302-95-4 | |
| MFCD00064139 | |
| [Na+].[H][C@@]12CC[C@H]([C@H](C)CCC([O-])=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])CC[C@]2([H])C[C@H](O)CC[C@]12C | |
| 414.56 | |
| ≥99% (TLC) |