missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Deoxycholic Acid, Sodium Salt, ≥99%, ULTROL™ Grade, Calbiochem™,
Supplier: MilliporeSigma™ 26410325GM
| Quantity | 25 g |
|---|---|
| Packaging | Glass Bottle |
Chemical Identifiers
| 302-95-4 | |
| 414.56 | |
| FHHPUSMSKHSNKW-SMOYURAASA-M | |
| sodium (4R)-4-[(1R,3aS,3bR,5aR,7R,9aS,9bS,11S,11aR)-7,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate |
| C24H39NaO4 | |
| MFCD00064139 | |
| 91810855 | |
| [Na+].[H][C@@]12CC[C@H]([C@H](C)CCC([O-])=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])CC[C@]2([H])C[C@H](O)CC[C@]12C |
Specifications
| White | |
| 25 g | |
| C24H39NaO4 | |
| FHHPUSMSKHSNKW-SMOYURAASA-M | |
| sodium (4R)-4-[(1R,3aS,3bR,5aR,7R,9aS,9bS,11S,11aR)-7,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate | |
| 91810855 | |
| ULTROL |
| 302-95-4 | |
| Glass Bottle | |
| MFCD00064139 | |
| [Na+].[H][C@@]12CC[C@H]([C@H](C)CCC([O-])=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])CC[C@]2([H])C[C@H](O)CC[C@]12C | |
| 414.56 | |
| ≥99% (TLC) |