missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Deoxycholic Acid, Sodium Salt, ≥99%, ULTROL™ Grade, Calbiochem™,

Supplier:  MilliporeSigma™ 2641035GM

 View more versions of this product

Catalog No. 2641035GM


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

White
5 g
C24H39NaO4
FHHPUSMSKHSNKW-SMOYURAASA-M
sodium (4R)-4-[(1R,3aS,3bR,5aR,7R,9aS,9bS,11S,11aR)-7,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate
91810855
ULTROL
302-95-4
Glass Bottle
MFCD00064139
[Na+].[H][C@@]12CC[C@H]([C@H](C)CCC([O-])=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])CC[C@]2([H])C[C@H](O)CC[C@]12C
414.56
≥99% (TLC)

Chemical Identifiers

302-95-4
414.56
FHHPUSMSKHSNKW-SMOYURAASA-M
sodium (4R)-4-[(1R,3aS,3bR,5aR,7R,9aS,9bS,11S,11aR)-7,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate
C24H39NaO4
MFCD00064139
91810855
[Na+].[H][C@@]12CC[C@H]([C@H](C)CCC([O-])=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])CC[C@]2([H])C[C@H](O)CC[C@]12C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions