missing translation for 'onlineSavingsMsg'
Learn More

Density Standard 998 kg/m3 HD Fitzgerald Ltd. Quality, MilliporeSigma™ Supelco™

Catalog No. 111015501
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015501 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015501 Supplier Merck Emd Millipore Supplier No. 1215610MLF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Traceable liquid density standard for calibration of laboratory density meters, supplied with an ISO 17025 accredited calibration certificate.

Featured Industries: Cleaning Products, Cosmetics, Flavors and Fragrances, Food and Beverages, Personal Care and Pharmaceutical (small molecule)

Specifications

Chemical Name or Material Water
CAS 7732-18-5
Density 1.000 g/mL (at 3.98°C)
Linear Formula H2O
MDL Number MFCD00011332
Boiling Point 100°C
Synonym Water
Type Density Standard
Quantity 10 mL
Formula Weight 18.02
Grade H&D; Fitzgerald Ltd. Quality
Show More Show Less