missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO Density Kit for Standard and Advanced Balances

Catalog No. 1814238
Change view
Click to view available options
Type:
Density Determination Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
1814238 Density Determination Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1814238 Supplier METTLER TOLEDO Supplier No. 30706714*

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Density kit for determining the density of solid samples, compatible with MX, MR, and MA series Standard and Advanced balances.

  • Converts balances into density measurement systems for solid samples
  • Replaces the weighing pan with a dedicated density kit setup
  • Compatible with MX, MR, and MA series balances
  • Supports readability levels of 0.01 mg, 0.1 mg, and 1 mg
  • Reliable accessory for expanding laboratory balance capabilities

Specifications

For Use With (Equipment) MX, MR, MA Balances
Includes Beaker, Adapters, Bracket, Wetting Agent, Thermometer
Type Density Determination Kit