missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Density Cube with Hook

Convenient for use during density, specific heat, and specific gravity investigations.

Supplier:  Eisco™ PH0108J

 View more versions of this product

Catalog No. S27353


Only null left

Description

Description

  • Designed for use in an educational setting
  • Not fabricated using ultra high precision machining
  • Cube features 0.75 in. (20 mm) sides with a 0.75 x 0.5 mm hook
  • Expect tolerances of ± 5%
Specifications

Specifications

Density, Specific Heat, and Specific Gravity Investigations
Hook (0.75 x 0.5 mm)
0.75 in.
0.75 in.
Density Cube
Brass
20 mm
20 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions