Learn More
Thermo Scientific™ Dense Storage Racks for Chest ULTs and Cryo Storage
A convenient, streamlined process to maximize sample storage. Our freezer racking and tubing solutions are designed to hold tubes ranging from 200μL to 5.0μL.
Supplier:  Thermo Scientific™ 1950834
 View more versions of this product
Catalog No. 11680087
Only null left
Description
Description
A convenient, streamlined process to maximize sample storage in your laboratory. Thermo Scientific™ Dense Storage Racks for Chest ULTs and Cryo Storage are designed to hold tubes ranging from 200μL up to 5.0μL, offering maximized sample storage capacity for ultra-low chest freezers and large-capacity cryogenic freezers.
  • Warranty varies by country, please contact us for details
Specifications
Chemical Identifiers
Racks for Tubes (cryogenic and ultra-low chest freezers)
Rectangular
3.6 in.
Tube Rack (Cryogenic and Ultra-Low Chest Freezers)
Silver
5.3 in.
9.3 cm
13.5 cm
Specifications
Racks for Tubes (cryogenic and ultra-low chest freezers)
Plates (9/Rack)
Matrix 1 mL Screw-top Tubes
5.3 in.
3.6 in.
Tube Rack (Cryogenic and Ultra-Low Chest Freezers)
13.5 cm
Silver
Rectangular
Thermo Scientific CryoPlus 1 (hold 7 racks), CryoPlus 2 (holds 16 racks), CryoPlus 3 (holds 30 racks), CryoPlus 4 (holds 51 racks)
Thermo Scientific mechanical cryogenic chest freezer 6.8 cu. ft. (holds 18 racks), 10.3 cu. ft. (holds 27 racks)
22.5 in.
9.3 cm
57.2 cm
Product Suggestions
Product Suggestions