missing translation for 'onlineSavingsMsg'
Learn More

Velp™ Denitrification Glass Bottle for RESPIROMETRIC Sensor

Catalog No. 11392148
Change view
Click to view available options
For Use With (Equipment):
RESPIROMETRIC Sensor
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
11392148 RESPIROMETRIC Sensor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11392148 Supplier Velp™ Supplier No. A00000410

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Denitrification glass bottle for use with the RESPIROMETRIC Sensor.

  • Bottle for the denitrification process

Specifications

Components Measured Nitric Oxide
For Use With (Equipment) RESPIROMETRIC Sensor
Type Gas Sensor
For Use With (Application) Denitrificaton