missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Dengue Virus Type 2 Monoclonal Antibody (3H5.1)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA183114
Description
This antibody is suitable for use in hemagglutination-inhibition tests and plaque-reduction assays. Mouse anti Dengue virus type 2 antibody, clone 3H5.1 recognizes Dengue virus type 2, a flavivirus known to cause acute febrile disease (haemorrhagic fevers) in humans.
Dengue fever and dengue hemorrhagic fever are acute febrile diseases, found in the tropics and Africa. Caused by one of four closely related virus serotypes of the genus Flavivirus, family Flaviviridae, each serotype is sufficiently different that there is no cross-protection and epidemics caused by multiple serotypes (hyperendemicity) can occur. Dengue is transmitted to humans by the Aedes aegypti (rarely Aedes albopictus) mosquito, which feeds during the day.Chemical Identifiers
| Dengue Virus Type 2 | |
| 1 mg/mL | |
| Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 0.1% sodium azide | |
| Native Dengue type 2 antigen (New Guinea C). | |
| Antibody |
| 3H5.1 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG1 | |
| Virus | |
| Dengue Virus 2; DENV; DENV2 | |
| Primary | |
| Protein G |
Specifications
| Dengue Virus Type 2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Mouse | |
| Protein G | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 |
| Immunocytochemistry | |
| 3H5.1 | |
| Unconjugated | |
| Dengue Virus 2; DENV; DENV2 | |
| Native Dengue type 2 antigen (New Guinea C). | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |