missing translation for 'onlineSavingsMsg'
Learn More

Denatonium Benzoate 98.0+%, TCI America™

Catalog No. p-7127120
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21245G 5 g
D212425G 25 g
D2124100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D21245G Supplier TCI America Supplier No. D21245G

Chemical Identifiers

CAS 3734-33-6
Molecular Formula C28H34N2O3
Molecular Weight (g/mol) 446.59
MDL Number MFCD00031578
InChI Key VWTINHYPRWEBQY-UHFFFAOYSA-N
Synonym denatonium benzoate, bitrex, lidocaine benzyl benzoate, anispray, aversion, gori, caswell no. 083bb, unii-m5ba6gaf1o, denatonium benzoate anhydrous
PubChem CID 19518
IUPAC Name benzyl({[(2,6-dimethylphenyl)carbamoyl]methyl})diethylazanium benzoate
SMILES [O-]C(=O)C1=CC=CC=C1.CC[N+](CC)(CC(=O)NC1=C(C)C=CC=C1C)CC1=CC=CC=C1

Specifications

Melting Point 163°C
Color White
Quantity 5 g
Formula Weight 446.59
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Denatonium Benzoate
EINECSNumber (3)-2724
RTECSNumber BO6650000
TSCA Yes